Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197935.1 | NP_922917.1 | 6 - 7 | 1121 | 62 | |
XM_009292744.3 | XP_009291019.1 | 7 - 8 | 1493 | 62 | ||
GenBank mRNA | AY333791.1 | - | 6 - 7 | 1121 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Danio rerio Probe spans exons |