Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: figla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198919.2 | NP_944601.2 | 2 - 3 | 527 | 85 | |
GenBank mRNA | AY302567.1 | - | 2 - 3 | 381 | 85 | |
BC092676.1 | - | 2 - 3 | 527 | 85 | ||
BC164317.1 | - | 2 - 3 | 415 | 85 | ||
EU723768.1 | - | 2 - 3 | 473 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |