Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199748.1 | NP_956042.1 | 5 - 6 | 614 | 150 | |
XM_005158720.4 | XP_005158777.1 | 5 - 6 | 823 | 150 | ||
GenBank mRNA | BC053169.1 | - | 5 - 6 | 614 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |