Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201005.1 | NP_957299.1 | 3 - 4 | 312 | 113 | |
GenBank mRNA | BC047847.1 | - | 3 - 4 | 312 | 113 | |
BC075751.1 | - | 3 - 4 | 276 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |