Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213387.1 | NP_998552.1 | 3 - 4 | 340 | 90 | |
GenBank mRNA | BC046026.1 | - | 3 - 4 | 340 | 90 | |
BC065888.1 | - | 3 - 4 | 310 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |