Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bves | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001847.2 | NP_001001847.2 | 5 - 6 | 775 | 98 | |
NM_001257162.1 | NP_001244091.1 | 5 - 6 | 748 | 98 | ||
NM_001257163.1 | NP_001244092.1 | 5 - 6 | 775 | 98 | ||
NM_001257164.1 | NP_001244093.1 | 5 - 6 | 748 | 98 | ||
GenBank mRNA | AY293117.1 | - | 5 - 6 | 773 | 98 | |
AY293118.1 | - | 5 - 6 | 746 | 98 | ||
BC086956.1 | - | 5 - 6 | 748 | 98 | ||
JN400527.1 | - | 5 - 6 | 773 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |