Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002749.2 | NP_001002749.1 | 5 - 6 | 1168 | 77 | |
GenBank mRNA | BC076406.1 | - | 5 - 6 | 1136 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |