Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: g6pca.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003512.2 | NP_001003512.2 | 3 - 4 | 439 | 65 | |
XM_009306434.3 | XP_009304709.1 | 3 - 4 | 687 | 65 | ||
GenBank mRNA | BC076446.1 | - | 3 - 4 | 363 | 65 | |
BC164161.1 | - | 2 - 3 | 363 | 65 | ||
EH484431.1 | - | 3 - 4 | 465 | 65 | ||
EH488933.1 | - | 3 - 4 | 445 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |