Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006105.1 | NP_001006105.1 | 2 - 3 | 607 | 81 | |
XM_005165301.4 | XP_005165358.1 | 3 - 4 | 704 | 81 | ||
GenBank mRNA | BC083206.1 | - | 2 - 3 | 607 | 81 | |
BC095205.1 | - | 3 - 4 | 676 | 81 | ||
BC164056.1 | - | 3 - 4 | 498 | 81 | ||
BC165929.1 | - | 2 - 3 | 366 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |