Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: matn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012385.1 | NP_001012385.1 | 5 - 6 | 1432 | 87 | |
XM_005156825.4 | XP_005156882.1 | 4 - 5 | 1346 | 87 | ||
XM_005156826.4 | XP_005156883.1 | 4 - 5 | 1063 | 87 | ||
XM_005156827.4 | XP_005156884.1 | 3 - 4 | 936 | 87 | ||
GenBank mRNA | AJ698458.1 | - | 4 - 5 | 1300 | 87 | |
AJ698459.1 | - | 3 - 4 | 916 | 87 | ||
AJ698730.2 | - | 4 - 5 | 1048 | 87 | ||
AJ717297.1 | - | 5 - 6 | 1432 | 87 | ||
BC107521.1 | - | 3 - 4 | 923 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |