Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020480.1 | NP_001018316.1 | 8 - 9 | 974 | 59 | |
GenBank mRNA | AB194986.1 | - | 8 - 9 | 989 | 59 | |
BC095383.1 | - | 8 - 9 | 974 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |