Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020639.1 | NP_001018475.1 | 7 - 8 | 887 | 72 | |
GenBank mRNA | BC095635.1 | - | 7 - 8 | 887 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |