Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: necab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328213.1 | NP_001315142.1 | 7 - 8 | 849 | 68 | |
XM_009293572.3 | XP_009291847.1 | 7 - 8 | 884 | 68 | ||
GenBank mRNA | BC098611.1 | - | 7 - 8 | 831 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |