Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abcg2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039639.2 | NP_001034728.2 | 13 - 14 | 1668 | 69 | |
XM_005156523.4 | XP_005156580.1 | 13 - 14 | 69 | |||
XM_009307089.3 | XP_009305364.1 | 13 - 14 | 69 | |||
GenBank mRNA | BC124086.1 | - | 13 - 14 | 1668 | 69 | |
BC165778.1 | - | 12 - 13 | 1647 | 69 | ||
DQ223860.1 | - | 12 - 13 | 1613 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |