Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfsf10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131843.2 | NP_571918.2 | 2 - 3 | 392 | 136 | |
XM_005172247.4 | XP_005172304.1 | 2 - 3 | 597 | 133 | ||
GenBank mRNA | AF250041.1 | - | 1 - 2 | 145 | 136 | |
BC044336.1 | - | 2 - 3 | 392 | 136 | ||
DQ812112.1 | - | 2 - 3 | 289 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |