Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meis1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131893.2 | NP_571968.1 | 6 - 7 | 1258 | 96 | |
GenBank mRNA | AF375871.2 | - | 6 - 7 | 1005 | 96 | |
BC060891.1 | - | 6 - 7 | 781 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |