Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fabp10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152960.1 | NP_694492.1 | 3 - 4 | 338 | 94 | |
GenBank mRNA | AF254642.2 | - | 3 - 4 | 338 | 94 | |
BC076219.1 | - | 3 - 4 | 353 | 94 | ||
BC081518.1 | - | 3 - 4 | 351 | 94 | ||
BC164928.1 | - | 3 - 4 | 360 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |