Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arntl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178300.1 | NP_840085.1 | 6 - 7 | 464 | 66 | |
XM_005166616.4 | XP_005166673.1 | 9 - 10 | 722 | 66 | ||
XM_009303573.3 | XP_009301848.1 | 8 - 9 | 653 | 66 | ||
XM_021477412.1 | XP_021333087.1 | 8 - 9 | 660 | 66 | ||
GenBank mRNA | AB087257.1 | - | 6 - 7 | 464 | 66 | |
BC134895.1 | - | 9 - 10 | 707 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |