Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r5ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194415.2 | NP_919396.1 | 6 - 7 | 1134 | 82 | |
GenBank mRNA | AY150214.1 | - | 5 - 6 | 681 | 82 | |
BC044398.1 | - | 6 - 7 | 950 | 82 | ||
BC078645.1 | - | 6 - 7 | 1134 | 82 | ||
BC165347.1 | - | 5 - 6 | 715 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |