Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199666.1 | NP_955960.1 | 3 - 4 | 409 | 83 | |
XM_005173100.4 | XP_005173157.1 | 3 - 4 | 457 | 83 | ||
GenBank mRNA | BC044423.1 | - | 3 - 4 | 409 | 83 | |
BC165021.1 | - | 3 - 4 | 284 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |