Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212584.1 | NP_997749.1 | 2 - 3 | 260 | 95 | |
XM_005165268.2 | XP_005165325.1 | 3 - 4 | 398 | 95 | ||
XM_021476007.1 | XP_021331682.1 | 3 - 4 | 607 | 95 | ||
GenBank mRNA | AF498292.1 | - | 1 - 2 | 170 | 95 | |
BC066490.1 | - | 2 - 3 | 260 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |