Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212859.2 | NP_998024.2 | 1 - 2 | 276 | 63 | |
GenBank mRNA | AY427649.1 | - | 1 - 2 | 179 | 63 | |
BC124141.1 | - | 1 - 2 | 276 | 63 | ||
BC165066.1 | - | 1 - 2 | 213 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |