Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003572.1 | NP_001003572.1 | 10 - 11 | 1069 | 119 | |
GenBank mRNA | BC077118.1 | - | 10 - 11 | 1069 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |