Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osbpl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004646.1 | NP_001004646.1 | 17 - 18 | 1884 | 154 | |
XM_005170028.4 | XP_005170085.1 | 17 - 18 | 2138 | 154 | ||
GenBank mRNA | BC078429.1 | - | 17 - 18 | 1884 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |