Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl9l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005575.1 | NP_001005575.1 | 3 - 4 | 731 | 59 | |
XM_005159238.4 | XP_005159295.1 | 3 - 4 | 835 | 59 | ||
GenBank mRNA | AY296057.1 | - | 3 - 4 | 731 | 59 | |
BC163544.1 | - | 3 - 4 | 711 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |