Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcsha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013457.1 | NP_001013475.1 | 4 - 5 | 459 | 68 | |
GenBank mRNA | BC091548.1 | - | 4 - 5 | 459 | 68 | |
BC164865.1 | - | 4 - 5 | 457 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |