Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cntn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014814.1 | NP_001014814.1 | 11 - 12 | 1467 | 82 | |
GenBank mRNA | AY301281.1 | - | 11 - 12 | 1467 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Danio rerio Probe spans exons |