Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grhl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001326533.1 | NP_001313462.1 | 3 - 4 | 520 | 106 | |
GenBank mRNA | BC096802.1 | - | 3 - 4 | 472 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |