Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smyd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030223.1 | NP_001025394.1 | 3 - 4 | 360 | 144 | |
XM_005164423.4 | XP_005164480.1 | 3 - 4 | 730 | 144 | ||
GenBank mRNA | DQ851822.1 | - | 3 - 4 | 201 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Probe spans exons |