Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgfbr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037683.2 | NP_001032772.2 | 2 - 3 | 352 | 86 | |
GenBank mRNA | BC109402.1 | - | 2 - 3 | 361 | 86 | |
EF462417.1 | - | 2 - 3 | 352 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |