Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: illr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040039.1 | NP_001035128.1 | 6 - 7 | 483 | 82 | |
NM_001044742.1 | NP_001038207.1 | 5 - 6 | 384 | 82 | ||
XM_021468134.1 | XP_021323809.1 | 6 - 7 | 510 | 82 | ||
GenBank mRNA | AY986756.1 | - | 6 - 7 | 483 | 82 | |
AY986757.1 | - | 5 - 6 | 384 | 82 | ||
BC163143.1 | - | 5 - 6 | 384 | 82 | ||
BC163144.1 | - | 5 - 6 | 384 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |