Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kti12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126418.1 | NP_001119890.1 | 1 - 2 | 261 | 109 | |
GenBank mRNA | BC122265.1 | - | 1 - 2 | 251 | 109 | |
BC164624.1 | - | 1 - 2 | 186 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |