Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: borcs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098247.2 | NP_001091717.1 | 4 - 5 | 363 | 77 | |
GenBank mRNA | BC162286.1 | - | 4 - 5 | 363 | 77 | |
BC162309.1 | - | 4 - 5 | 363 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |