Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320073.1 | NP_001307002.1 | 3 - 4 | 314 | 76 | |
NM_131758.2 | NP_571833.1 | 2 - 3 | 284 | 76 | ||
XM_017352833.2 | XP_017208322.1 | 4 - 5 | 372 | 76 | ||
XM_017352834.2 | XP_017208323.1 | 3 - 4 | 342 | 76 | ||
GenBank mRNA | BC091908.1 | - | 3 - 4 | 279 | 76 | |
BC165270.1 | - | 3 - 4 | 203 | 76 | ||
Y13473.1 | - | 283 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |