Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144764.2 | NP_658910.2 | 5 - 6 | 588 | 95 | |
GenBank mRNA | AJ306396.1 | - | 5 - 6 | 591 | 95 | |
BC121194.1 | - | 5 - 6 | 532 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |