Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328433.1 | NP_001315362.1 | 3 - 4 | 815 | 80 | |
XM_021478599.1 | XP_021334274.1 | 3 - 4 | 667 | 80 | ||
GenBank mRNA | AF448504.1 | - | 3 - 4 | 781 | 80 | |
BC128812.1 | - | 3 - 4 | 777 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |