Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpi1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153667.2 | NP_705953.1 | 5 - 6 | 621 | 88 | |
GenBank mRNA | AF387820.1 | - | 5 - 6 | 621 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |