Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkcbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199641.1 | NP_955935.1 | 11 - 12 | 1578 | 96 | |
XM_005161943.4 | XP_005162000.1 | 12 - 13 | 1687 | 96 | ||
XM_005161944.4 | XP_005162001.1 | 12 - 13 | 2149 | 96 | ||
XM_005161945.4 | XP_005162002.1 | 12 - 13 | 1992 | 96 | ||
XM_005161946.4 | XP_005162003.1 | 12 - 13 | 1608 | 96 | ||
XM_009296662.3 | XP_009294937.1 | 12 - 13 | 1587 | 96 | ||
XM_009296664.3 | XP_009294939.1 | 12 - 13 | 1590 | 96 | ||
XM_009296665.3 | XP_009294940.1 | 12 - 13 | 1687 | 96 | ||
GenBank mRNA | BC058064.1 | - | 11 - 12 | 1578 | 96 | |
BC091798.1 | - | 12 - 13 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |