Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndel1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201344.1 | NP_958501.1 | 4 - 5 | 496 | 94 | |
XM_009306604.3 | XP_009304879.1 | 4 - 5 | 522 | 94 | ||
XM_009306605.2 | XP_009304880.1 | 4 - 5 | 522 | 94 | ||
GenBank mRNA | BC055587.1 | - | 4 - 5 | 496 | 94 | |
BC164385.1 | - | 3 - 4 | 423 | 94 | ||
EF133514.1 | - | 4 - 5 | 424 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |