Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201501.1 | NP_958909.1 | 2 - 3 | 504 | 74 | |
GenBank mRNA | BC045466.1 | - | 2 - 3 | 504 | 74 | |
BC065634.1 | - | 2 - 3 | 459 | 74 | ||
BC165047.1 | - | 1 - 2 | 208 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |