Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201501.1 | NP_958909.1 | 5 - 6 | 870 | 63 | |
GenBank mRNA | BC045466.1 | - | 5 - 6 | 870 | 63 | |
BC065634.1 | - | 5 - 6 | 825 | 63 | ||
BC165047.1 | - | 4 - 5 | 574 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |