Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adh8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203460.1 | NP_982285.1 | 8 - 9 | 1120 | 97 | |
XM_021480549.1 | XP_021336224.1 | 8 - 9 | 1242 | 97 | ||
GenBank mRNA | AY309075.1 | - | 8 - 9 | 1120 | 97 | |
BC071367.1 | - | 8 - 9 | 1117 | 97 | ||
BC154296.1 | - | 7 - 8 | 975 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |