Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002060.1 | NP_001002060.1 | 7 - 8 | 823 | 111 | |
XM_009302036.3 | XP_009300311.1 | 7 - 8 | 930 | 111 | ||
GenBank mRNA | BC071346.1 | - | 7 - 8 | 823 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |