Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002173.1 | NP_001002173.1 | 6 - 7 | 1552 | 96 | |
GenBank mRNA | AY542372.2 | - | 6 - 7 | 1567 | 96 | |
BC074092.1 | - | 6 - 7 | 1552 | 96 | ||
BC164696.1 | - | 5 - 6 | 1508 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |