Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002437.1 | NP_001002437.1 | 2 - 3 | 293 | 78 | |
XM_021466676.1 | XP_021322351.1 | 2 - 3 | 217 | 78 | ||
GenBank mRNA | BC076127.1 | - | 2 - 3 | 293 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |