Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: camk2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002542.1 | NP_001002542.1 | 13 - 14 | 1241 | 71 | |
XM_009306662.3 | XP_009304937.1 | 13 - 14 | 1284 | 71 | ||
XM_021471546.1 | XP_021327221.1 | 13 - 14 | 1283 | 71 | ||
GenBank mRNA | BC076266.1 | - | 13 - 14 | 1241 | 71 | |
BC154768.1 | - | 13 - 14 | 1196 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |