Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a36a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002667.1 | NP_001002667.1 | 5 - 6 | 816 | 73 | |
XM_009298709.3 | XP_009296984.1 | 2 - 3 | 3131 | 73 | ||
XM_009298710.3 | XP_009296985.1 | 3 - 4 | 3014 | 73 | ||
XM_021479882.1 | XP_021335557.1 | 6 - 7 | 940 | 73 | ||
XM_021479889.1 | XP_021335564.1 | 3 - 4 | 2992 | 73 | ||
GenBank mRNA | BC076521.1 | - | 5 - 6 | 816 | 73 | |
BC164094.1 | - | 5 - 6 | 488 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |