Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004110.1 | NP_001004110.1 | 19 - 20 | 1994 | 122 | |
XM_005158836.4 | XP_005158893.1 | 20 - 21 | 2248 | 122 | ||
XM_009293188.3 | XP_009291463.1 | 19 - 20 | 2239 | 122 | ||
XM_021467147.1 | XP_021322822.1 | 20 - 21 | 2252 | 122 | ||
XM_021467148.1 | XP_021322823.1 | 19 - 20 | 2242 | 122 | ||
GenBank mRNA | AY618701.1 | - | 19 - 20 | 1994 | 122 | |
AY691328.1 | - | 20 - 21 | 2147 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |