Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007413.1 | NP_001007414.1 | 4 | 1302 | 104 | |
GenBank mRNA | BC085372.1 | - | 4 | 1302 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |