Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 14 - 15 | 1699 | 113 | |
XM_005160684.4 | XP_005160741.1 | 13 - 14 | 1330 | 113 | ||
XM_009294709.3 | XP_009292984.1 | 14 - 15 | 1724 | 113 | ||
GenBank mRNA | DT075242.1 | - | 3 - 4 | 296 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |